Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PABPC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135653.1 | NP_001129125.1 | 14 | 2688 | ||
NM_001135654.1 | NP_001129126.1 | 13 | 2601 | |||
NM_003819.3 | NP_003810.1 | 14 | 2640 | |||
GenBank mRNA | AB208962.1 | - | 8 | 884 | ||
AK091110.1 | - | 2 | 1744 | |||
AK125926.1 | - | 2 | 2232 | |||
AK222917.1 | - | 8 | 925 | |||
BC029969.1 | - | 14 | 2240 | |||
BC065540.1 | - | 13 | 2504 | 6061561 | ||
BC071591.1 | - | 14 | 2626 | 5265829 | ||
BC094755.1 | - | 14 | 2629 | 30331896 | ||
U33818.1 | - | 14 | 1895 | |||
U75686.1 | - | 14 | 2536 |