Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100505502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011519798.2 | XP_011518100.1 | 1 | 1198 | ||
GenBank mRNA | AK094743.1 | - | 2 | 1297 |