Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLNC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127487.1 | NP_001120959.1 | 4 | 957 | ||
NM_001458.4 | NP_001449.3 | 4 | 957 | |||
GenBank mRNA | AB371585.1 | - | 4 | 977 | ||
AF089841.1 | - | 4 | 797 | |||
AF146692.1 | - | 4 | 960 | |||
AJ012737.1 | - | 4 | 766 |