Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102723532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011546215.2 | XP_011544517.1 | 1 | 21 |
Gene Symbol: LOC105369274 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011543799.2 | XP_011542101.1 | 1 | 15 |
Gene Symbol: OR4M1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005500.1 | NP_001005500.1 | 1 | 588 | ||
XM_017021338.1 | XP_016876827.1 | 1 | 1247 | |||
XM_017021339.1 | XP_016876828.1 | 1 | 1247 | |||
GenBank mRNA | BC136987.1 | - | 1 | 669 | 9020979 |
Gene Symbol: OR4M2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004719.2 | NP_001004719.2 | 1 | 686 | ||
GenBank mRNA | BC136985.1 | - | 1 | 686 | 9020977 |