Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144628.3 | NP_653229.1 | 4, 5 | 681 | ||
NR_111901.1 | 4, 5 | 681 | ||||
XM_005260661.1 | XP_005260718.1 | 4, 5 | 671 | |||
XM_006723540.3 | XP_006723603.1 | 3, 4 | 460 | |||
XM_017027645.1 | XP_016883134.1 | 3, 4 | 452 | |||
GenBank mRNA | AB449906.1 | - | 4, 5 | 533 | ||
AK095110.1 | - | 4 | 840 | |||
AK127062.1 | - | 4, 5 | 661 | |||
AK291648.1 | - | 4, 5 | 644 | |||
BC014983.1 | - | 4, 5 | 631 | 4861869 | ||
BX647370.1 | - | 1 | 212 | |||
DA230581.1 | - | 1, 2 | 194 |