Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144628.3 | NP_653229.1 | 4 | 644 | ||
NR_111901.1 | 4 | 644 | ||||
XM_005260661.1 | XP_005260718.1 | 4 | 634 | |||
XM_006723540.3 | XP_006723603.1 | 3 | 423 | |||
XM_017027645.1 | XP_016883134.1 | 3 | 415 | |||
GenBank mRNA | AB449906.1 | - | 4 | 496 | ||
AK095110.1 | - | 4 | 636 | |||
AK127062.1 | - | 4 | 624 | |||
AK291648.1 | - | 4 | 607 | |||
BC014983.1 | - | 4 | 594 | 4861869 | ||
DA230581.1 | - | 1 | 157 |