Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PABPN1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080487.2 | NP_001073956.2 | 4, 5 | 623 | ||
XM_017023230.1 | XP_016878719.1 | 6, 7 | 1878 | |||
GenBank mRNA | EU190484.1 | - | 4, 5 | 581 |