Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000136.2 | NP_000127.2 | 12 | 1397 | ||
NM_001243743.1 | NP_001230672.1 | 12 | 1339 | |||
NM_001243744.1 | NP_001230673.1 | 12 | 1397 | |||
XM_005251802.3 | XP_005251859.1 | 6 | 1499 | |||
XM_006717001.2 | XP_006717064.1 | 11 | 1214 | |||
XM_006717002.3 | XP_006717065.1 | 12 | 1379 | |||
XM_006717004.3 | XP_006717067.1 | 11 | 1303 | |||
XM_011518365.2 | XP_011516667.1 | 12 | 1372 | |||
XM_011518366.2 | XP_011516668.1 | 12 | 1379 | |||
XM_011518367.2 | XP_011516669.1 | 13 | 1509 | |||
XM_017014452.1 | XP_016869941.1 | 13 | 1524 | |||
XM_017014453.1 | XP_016869942.1 | 13 | 1471 | |||
XM_017014454.1 | XP_016869943.1 | 12 | 1398 | |||
GenBank mRNA | AK222871.1 | - | 12 | 1359 | ||
AK312548.1 | - | 12 | 1262 | |||
BC015748.1 | - | 12 | 1387 | 4864916 | ||
KU178051.1 | - | 11 | 1135 | |||
KU178052.1 | - | 12 | 1280 | |||
X66893.1 | - | 12 | 1390 | |||
X66894.1 | - | 12 | 1308 |