Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000137.2 | NP_000128.1 | 5 | 588 | ||
GenBank mRNA | AK297965.1 | - | 6 | 570 | ||
AK313951.1 | - | 5 | 583 | |||
BC002527.1 | - | 5 | 426 | 3139988 | ||
BQ225295.1 | - | 5 | 588 | |||
BT007160.1 | - | 5 | 424 | |||
BX537608.1 | - | 4 | 2453 | |||
M55150.1 | - | 5 | 480 | |||
X51728.1 | - | 4 | 351 |