Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006432.3 | NP_006423.1 | 3 | 419 | ||
XM_017020930.1 | XP_016876419.1 | 3 | 513 | |||
GenBank mRNA | AK222474.1 | - | 3 | 339 | ||
AK298975.1 | - | 3 | 319 | |||
AK300879.1 | - | 3 | 346 | |||
AK311931.1 | - | 3 | 380 | |||
BC002532.2 | - | 3 | 418 | 3140870 | ||
X67698.1 | - | 3 | 322 |