Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017882.2 | NP_060352.1 | 5 | 660 | ||
GenBank mRNA | AK000568.1 | - | 5 | 605 | ||
AK027604.1 | - | 5 | 628 | |||
AK074753.1 | - | 2 | 154 | |||
AK291175.1 | - | 5 | 587 | |||
AK293197.1 | - | 5 | 836 | |||
BC010849.1 | - | 5 | 574 | 3878776 | ||
BC013130.1 | - | 5 | 511 | 4517748 | ||
CR457244.1 | - | 5 | 502 | |||
KU178671.1 | - | 5 | 502 | |||
KU178672.1 | - | 5 | 621 | |||
KU178673.1 | - | 4 | 403 | |||
KU178674.1 | - | 4 | 313 |