Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017882.2 | NP_060352.1 | 7 | 1332 | ||
GenBank mRNA | AK000568.1 | - | 7 | 1273 | ||
AK027604.1 | - | 7 | 1296 | |||
AK074753.1 | - | 4 | 822 | |||
BC010849.1 | - | 7 | 1242 | 3878776 | ||
BC013130.1 | - | 7 | 1179 | 4517748 | ||
CN427564.1 | - | 1 | 98 |