Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDOB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000035.3 | NP_000026.2 | 2, 3 | 187 | ||
GenBank mRNA | AK290795.1 | - | 2, 3 | 181 | ||
BC029399.1 | - | 2, 3 | 181 | 4593670 | ||
X01098.1 | - | 2, 3 | 171 | |||
X02747.1 | - | 2, 3 | 230 |