Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATP7B SIRNA Assays ›
Gene Symbol:
ATP7B
Gene Name:
ATPase copper transporting beta
Gene Aliases:
PWD, WC1, WD, WND
Chromosome Location:
Chr.13: 51932669 - 52012130 on Build GRCh38
UniGene ID:
Hs.492280
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s529130
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATP7B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000053.3
NP_000044.2
16
3681
NM_001005918.2
NP_001005918.1
12
3060
NM_001243182.1
NP_001230111.1
17
3348
XM_005266423.2
XP_005266480.1
16
3586
XM_005266424.4
XP_005266481.1
16
4207
XM_005266427.2
XP_005266484.1
15
3509
XM_005266428.1
XP_005266485.1
14
3491
XM_005266430.4
XP_005266487.1
17
3689
XM_005266431.3
XP_005266488.1
16
3741
XM_005266432.2
XP_005266489.1
13
3257
XM_006719837.3
XP_006719900.1
16
3835
XM_006719838.1
XP_006719901.1
10
1476
XM_006719839.1
XP_006719902.1
9
1293
XM_011535117.2
XP_011533419.1
16
4762
XM_011535118.1
XP_011533420.1
15
3608
XM_011535119.1
XP_011533421.1
15
3560
XM_011535122.2
XP_011533424.1
15
2434
XM_017020627.1
XP_016876116.1
16
3585
XM_017020628.1
XP_016876117.1
15
2430
GenBank mRNA
AB209461.1
-
12
4270
AK302074.1
-
9
1302
BC117200.1
-
8
2308
40125751
BC143973.1
-
15
3403
9052491
BC143974.1
-
16
3382
BC143975.1
-
15
3364
9052493
BC143976.1
-
16
3454
9052494
BX648261.1
-
9
2872
DQ015922.1
-
17
3191
L25591.1
-
11
2458
U03464.1
-
16
3524
U11700.1
-
16
3686
Back To Top
More Information
Panther Classification:
Molecular Function -
primary active transporter
Gene Ontology Categories:
Function(s)
Process(es)
copper ion transport
cellular copper ion homeostasis
cellular zinc ion homeostasis
lactation
circadian rhythm
response to zinc ion
copper ion import
intracellular copper ion transport
ion transmembrane transport
response to copper ion
sequestering of calcium ion
response to cAMP
copper ion export
copper-exporting ATPase activity
copper ion binding
protein binding
ATP binding
zinc ion binding
copper-transporting ATPase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-wh4bk:80/10.239.163.60:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0