Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRRS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013660.2 | NP_001013682.2 | 6 | 1164 | ||
XM_005270861.3 | XP_005270918.1 | 6 | 691 | |||
XM_011541451.2 | XP_011539753.1 | 6 | 1164 | |||
XM_011541452.1 | XP_011539754.1 | 6 | 691 | |||
XM_017001271.1 | XP_016856760.1 | 4 | 575 | |||
GenBank mRNA | AK131302.1 | - | 6 | 1164 |