Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EVC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306090.1 | NP_001293019.1 | 7 | 1098 | ||
NM_001306092.1 | NP_001293021.1 | 7 | 1098 | |||
NM_153717.2 | NP_714928.1 | 7 | 1098 | |||
XM_006713865.3 | XP_006713928.1 | 7 | 1095 | |||
XM_006713866.3 | XP_006713929.1 | 7 | 1095 | |||
XM_011513419.2 | XP_011511721.1 | 7 | 1092 | |||
XM_017007883.1 | XP_016863372.1 | 7 | 1092 | |||
GenBank mRNA | AF216184.1 | - | 7 | 1098 | ||
AF216185.1 | - | 7 | 1098 | |||
AF239742.1 | - | 7 | 914 | |||
AK025394.1 | - | 2 | 227 | |||
BC085608.1 | - | 7 | 1080 | 30523373 |