Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMPDH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000883.3 | NP_000874.2 | 9, 10 | 964 | ||
NM_001102605.1 | NP_001096075.1 | 8, 9 | 934 | |||
NM_001142573.1 | NP_001136045.1 | 6, 7 | 734 | |||
NM_001142574.1 | NP_001136046.1 | 6, 7 | 719 | |||
NM_001142575.1 | NP_001136047.1 | 5, 6 | 659 | |||
NM_001142576.1 | NP_001136048.1 | 8, 9 | 865 | |||
NM_001304521.1 | NP_001291450.1 | 6, 7 | 762 | |||
NM_183243.2 | NP_899066.1 | 7, 8 | 856 | |||
XM_017012168.1 | XP_016867657.1 | 8, 9 | 934 | |||
XM_017012169.1 | XP_016867658.1 | 7, 8 | 856 | |||
XM_017012170.1 | XP_016867659.1 | 6, 7 | 763 | |||
XM_017012171.1 | XP_016867660.1 | 8, 9 | 1052 | |||
XM_017012172.1 | XP_016867661.1 | 8, 9 | 1052 | |||
XM_017012173.1 | XP_016867662.1 | 8, 9 | 934 | |||
GenBank mRNA | AK054640.1 | - | 5, 6 | 631 | ||
AK054667.1 | - | 7, 8 | 856 | |||
AK092452.1 | - | 8, 9 | 901 | |||
AK122994.1 | - | 8, 9 | 933 | |||
AK131294.1 | - | 6, 7 | 681 | |||
AK293413.1 | - | 6, 7 | 691 | |||
AK297943.1 | - | 6, 7 | 706 | |||
BC033622.1 | - | 7, 8 | 847 | 5210847 | ||
CD014008.1 | - | 7, 8 | 648 | |||
CD014010.1 | - | 8, 9 | 770 | |||
CR933672.1 | - | 900 | ||||
J05272.1 | - | 9, 10 | 1228 |