Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000280.4 | NP_000271.1 | 8 | 1204 | ||
NM_001127612.1 | NP_001121084.1 | 8 | 1118 | |||
NM_001258462.1 | NP_001245391.1 | 9 | 1160 | |||
NM_001258463.1 | NP_001245392.1 | 9 | 1098 | |||
NM_001258464.1 | NP_001245393.1 | 8 | 1106 | |||
NM_001258465.1 | NP_001245394.1 | 7 | 1092 | |||
NM_001310158.1 | NP_001297087.1 | 9 | 1201 | |||
NM_001310159.1 | NP_001297088.1 | 6 | 777 | |||
NM_001310160.1 | NP_001297089.1 | 5 | 2415 | |||
NM_001310161.1 | NP_001297090.1 | 6 | 967 | |||
NM_001604.5 | NP_001595.2 | 9 | 1148 | |||
GenBank mRNA | AB209177.1 | - | 6 | 5556 | ||
AB593092.1 | - | 9 | 1099 | |||
AB593093.1 | - | 9 | 1098 | |||
AB593094.1 | - | 9 | 1201 | |||
AK074881.1 | - | 7 | 1539 | |||
AK094172.1 | - | 3 | 1842 | |||
AK094249.1 | - | 5 | 2225 | |||
AK314470.1 | - | 8 | 1096 | |||
AY047583.1 | - | 6 | 779 | |||
AY707088.1 | - | 4 | 649 | |||
BC011953.1 | - | 8 | 990 | 3880468 | ||
BX640762.1 | - | 9 | 1127 | |||
GQ141695.1 | - | 5 | 697 | |||
KP255960.1 | - | 5 | 691 | |||
M77844.1 | - | 7 | 1011 | |||
M93650.1 | - | 8 | 1066 |