Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037343.1 | NP_001032420.1 | 20, 21 | 3048 | ||
NM_003159.2 | NP_003150.1 | 19, 20 | 3052 | |||
GenBank mRNA | AY217744.1 | - | 19, 20 | 3020 | ||
Y15057.1 | - | 19, 20 | 3020 |