Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VEGFC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005429.4 | NP_005420.1 | 2 | 651 | ||
GenBank mRNA | AK313879.1 | - | 2 | 417 | ||
BC035212.1 | - | 2 | 482 | 4753457 | ||
BC063685.1 | - | 2 | 633 | 4513934 | ||
CR541897.1 | - | 2 | 217 | |||
U43142.1 | - | 2 | 588 | |||
U58111.1 | - | 2 | 540 | |||
X94216.1 | - | 2 | 568 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | X84838.1 | - | 1 | 80 | Chr. 4 - 176683542 - 176729747 [-] on Build GRCh38 |