Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018041.1 | NP_001018051.1 | 2 | 806 | ||
NM_005670.3 | NP_005661.1 | 2 | 806 | |||
XM_006715564.3 | XP_006715627.1 | 2 | 969 | |||
XM_011536113.2 | XP_011534415.1 | 2 | 969 | |||
XM_011536116.1 | XP_011534418.1 | 2 | 182 | |||
XM_017011300.1 | XP_016866789.1 | 1 | 148 | |||
GenBank mRNA | AF084535.2 | - | 2 | 449 | ||
AF284580.1 | - | 2 | 806 | |||
AF454491.1 | - | 2 | 227 | |||
AF454492.1 | - | 2 | 990 | |||
AF454493.1 | - | 3 | 694 | |||
AJ130763.1 | - | 2 | 208 | |||
AJ130764.1 | - | 2 | 208 | |||
AK299497.1 | - | 2 | 176 | |||
BC005286.1 | - | 2 | 445 | |||
BC070047.1 | - | 2 | 473 | 5287405 |