Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKKS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018848.3 | NP_061336.1 | 6 | 2250 | ||
NM_170784.2 | NP_740754.1 | 6 | 2140 | |||
NR_072977.1 | 5 | 738 | ||||
GenBank mRNA | AF221992.1 | - | 6 | 2095 | ||
AF221993.1 | - | 6 | 2247 | |||
AK025741.1 | - | 6 | 2075 | |||
AK291925.1 | - | 6 | 2089 | |||
AK296439.1 | - | 5 | 732 | |||
AL157427.1 | - | 6 | 2080 | |||
BC028973.2 | - | 6 | 2043 | 3921348 |