Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPF31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015629.3 | NP_056444.3 | 8 | 1184 | ||
XM_006723137.3 | XP_006723200.1 | 8 | 1094 | |||
GenBank mRNA | AB593024.1 | - | 9 | 2103 | ||
AB593025.1 | - | 8 | 840 | |||
AF308303.1 | - | 7 | 659 | |||
AK098547.1 | - | 8 | 823 | |||
AK301709.1 | - | 8 | 849 | |||
AL050369.1 | - | 8 | 823 | |||
AY040822.1 | - | 8 | 1145 | |||
BC117389.1 | - | 8 | 1108 | 40125940 |