Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HGSNAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152419.2 | NP_689632.2 | 2 | 216 | ||
XM_005273409.1 | XP_005273466.1 | 2 | 216 | |||
XM_005273410.1 | XP_005273467.1 | 2 | 216 | |||
XM_005273411.1 | XP_005273468.1 | 2 | 216 | |||
XM_005273412.3 | XP_005273469.1 | 2 | 216 | |||
XM_017013088.1 | XP_016868577.1 | 2 | 216 | |||
XM_017013089.1 | XP_016868578.1 | 2 | 120 | |||
GenBank mRNA | AK304441.1 | - | 2 | 168 | ||
BC042037.1 | - | 2 | 161 | |||
BE327420.1 | - | 1 | 64 | |||
DA395558.1 | - | 2 | 170 | |||
DB128369.1 | - | 2 | 216 |