Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152594.2 | NP_689807.1 | 3 | 634 | ||
XM_005254202.3 | XP_005254259.1 | 4 | 1434 | |||
XM_011521289.2 | XP_011519591.1 | 3 | 331 | |||
GenBank mRNA | AY299089.1 | - | 3 | 634 |