Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000038.5 | NP_000029.2 | 15 | 1997 | ||
NM_001127510.2 | NP_001120982.1 | 16 | 2105 | |||
NM_001127511.2 | NP_001120983.2 | 13 | 2071 | |||
GenBank mRNA | AB210001.1 | - | 16 | 2069 | ||
AK289567.1 | - | 15 | 2059 | |||
AK294544.1 | - | 13 | 2055 | |||
AK309033.1 | - | 2 | 266 | |||
M73548.1 | - | 14 | 1950 | |||
M74088.1 | - | 14 | 1930 |