Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002020.4 | NP_002011.2 | 7 | 962 | ||
NM_182925.4 | NP_891555.2 | 7 | 962 | |||
XM_011534478.2 | XP_011532780.1 | 7 | 1340 | |||
XM_011534484.2 | XP_011532786.1 | 5 | 520 | |||
XM_017009263.1 | XP_016864752.1 | 7 | 1346 | |||
XM_017009264.1 | XP_016864753.1 | 7 | 1346 | |||
XM_017009265.1 | XP_016864754.1 | 7 | 1347 | |||
XM_017009266.1 | XP_016864755.1 | 7 | 1341 | |||
XM_017009267.1 | XP_016864756.1 | 7 | 1341 | |||
XM_017009268.1 | XP_016864757.1 | 7 | 905 | |||
GenBank mRNA | AK291679.1 | - | 7 | 938 | ||
AK309910.1 | - | 7 | 938 | |||
AY233382.1 | - | 7 | 903 | |||
AY233383.1 | - | 7 | 903 | |||
EU826564.1 | - | 7 | 883 | |||
U43143.1 | - | 7 | 913 | |||
X68203.1 | - | 7 | 902 | |||
X69878.1 | - | 7 | 904 |