Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244808.1 | NP_001231737.1 | 21 | 3199 | ||
NM_001244810.1 | NP_001231739.1 | 21 | 3147 | |||
NM_001244812.1 | NP_001231741.1 | 16 | 2499 | |||
NM_001244813.1 | NP_001231742.1 | 15 | 2391 | |||
NM_001244814.1 | NP_001231743.1 | 17 | 2896 | |||
NM_001244815.1 | NP_001231744.1 | 16 | 2837 | |||
NM_001244816.1 | NP_001231745.1 | 20 | 3004 | |||
NM_032682.5 | NP_116071.2 | 21 | 3099 | |||
XM_006713102.2 | XP_006713165.1 | 20 | 3060 | |||
XM_011533584.2 | XP_011531886.1 | 21 | 3170 | |||
XM_011533585.2 | XP_011531887.1 | 20 | 3022 | |||
XM_017006165.1 | XP_016861654.1 | 20 | 2941 | |||
GenBank mRNA | AF250920.1 | - | 20 | 2997 |