Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CES2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003869.5 | NP_003860.2 | 7 | 2199 | ||
NM_198061.2 | NP_932327.1 | 7 | 2199 | |||
NR_036684.1 | 7 | 2437 | ||||
XM_011523421.2 | XP_011521723.1 | 7 | 1637 | |||
XM_017023818.1 | XP_016879307.1 | 7 | 1637 | |||
GenBank mRNA | AK095522.1 | - | 7 | 2437 | ||
AK222724.1 | - | 7 | 1077 | |||
AK290482.1 | - | 7 | 1479 | |||
AL713761.1 | - | 7 | 2152 | |||
BC032095.2 | - | 7 | 2183 | 4298532 | ||
BC071874.1 | - | 7 | 1138 | |||
BC098417.1 | - | 7 | 1141 | |||
BX538086.1 | - | 7 | 2200 | |||
D50579.1 | - | 7 | 1074 | |||
U60553.1 | - | 7 | 980 | |||
Y09616.1 | - | 7 | 1085 |