Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301347.1 | NP_001288276.1 | 4 | 646 | ||
NM_001460.4 | NP_001451.2 | 6 | 927 | |||
GenBank mRNA | AK304385.1 | - | 4 | 554 | ||
AK312877.1 | - | 6 | 833 | |||
AL833218.1 | - | 6 | 816 | |||
BC005894.1 | - | 6 | 833 | 4293031 | ||
BT006979.1 | - | 5 | 769 | |||
Y09267.1 | - | 6 | 829 |