Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002294.2 | NP_001002294.1 | 9 | 1667 | ||
NM_001319173.1 | NP_001306102.1 | 10 | 1777 | |||
NM_001319174.1 | NP_001306103.1 | 8 | 1478 | |||
NM_006894.5 | NP_008825.4 | 9 | 1650 | |||
XM_011509345.2 | XP_011507647.1 | 10 | 1798 | |||
GenBank mRNA | AK223166.1 | - | 9 | 1659 | ||
AK296105.1 | - | 8 | 1452 | |||
AK298406.1 | - | 10 | 1762 | |||
AK313197.1 | - | 9 | 1640 | |||
BC032016.1 | - | 9 | 1602 | 5175615 | ||
KU178077.1 | - | 8 | 1556 | |||
KU178078.1 | - | 5 | 666 | |||
M83772.1 | - | 9 | 1695 | |||
Z47552.1 | - | 9 | 1649 |