Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001557.2 | NP_001001557.1 | 2 | 1403 | ||
GenBank mRNA | AJ537424.1 | - | 2 | 1403 | ||
BC043222.2 | - | 1 | 543 | |||
BC127947.1 | - | 4 | 870 | 40132943 | ||
BC127948.1 | - | 3 | 1076 | 40132947 |