Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002022.2 | NP_002013.1 | 6 | 946 | ||
XM_005245045.3 | XP_005245102.1 | 5 | 857 | |||
XM_005245046.3 | XP_005245103.1 | 5 | 809 | |||
XM_011509347.2 | XP_011507649.1 | 6 | 998 | |||
XM_011509348.2 | XP_011507650.1 | 4 | 668 | |||
XM_011509349.2 | XP_011507651.1 | 6 | 996 | |||
GenBank mRNA | BC002780.2 | - | 6 | 895 | 3618884 | |
BT007444.1 | - | 4 | 565 | |||
Z11737.1 | - | 6 | 782 |