Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002022.2 | NP_002013.1 | 8 | 1512 | ||
XM_005245045.3 | XP_005245102.1 | 7 | 1423 | |||
XM_005245046.3 | XP_005245103.1 | 7 | 1375 | |||
XM_006711243.3 | XP_006711306.1 | 6 | 1033 | |||
XM_011509347.2 | XP_011507649.1 | 8 | 1564 | |||
XM_011509348.2 | XP_011507650.1 | 6 | 1234 | |||
GenBank mRNA | BC002780.2 | - | 8 | 1461 | 3618884 | |
BT007444.1 | - | 6 | 1131 | |||
Z11737.1 | - | 8 | 1348 |