Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000512.4 | NP_000503.1 | 5, 6 | 659 | ||
NM_001323543.1 | NP_001310472.1 | 4, 5 | 539 | |||
NM_001323544.1 | NP_001310473.1 | 6, 7 | 833 | |||
XM_005256301.3 | XP_005256358.1 | 5, 6 | 659 | |||
XM_011522982.2 | XP_011521284.1 | 6, 7 | 705 | |||
XM_017023111.1 | XP_016878600.1 | 5, 6 | 2556 | |||
XM_017023112.1 | XP_016878601.1 | 5, 6 | 2556 | |||
XM_017023113.1 | XP_016878602.1 | 4, 5 | 517 | |||
GenBank mRNA | AA436724.1 | - | 1, 2 | 157 | ||
AK131096.1 | - | 3, 4 | 3978 | |||
AK294390.1 | - | 3, 4 | 409 | |||
AK308269.1 | - | 6, 7 | 689 | |||
AK312655.1 | - | 5, 6 | 636 | |||
AY129021.1 | - | 3, 4 | 336 | |||
BC050684.1 | - | 5, 6 | 648 | 6091623 | ||
BC056151.1 | - | 5, 6 | 640 | 6026248 | ||
DA149503.1 | - | 4, 5 | 516 |