Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPGR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000328.2 | NP_000319.1 | 10 | 1296 | ||
NM_001034853.1 | NP_001030025.1 | 10 | 1296 | |||
XM_005272633.2 | XP_005272690.1 | 10 | 1325 | |||
XM_011543940.2 | XP_011542242.1 | 10 | 1292 | |||
XM_017029710.1 | XP_016885199.1 | 10 | 1325 | |||
XM_017029711.1 | XP_016885200.1 | 10 | 1322 | |||
XM_017029712.1 | XP_016885201.1 | 10 | 1322 | |||
GenBank mRNA | AJ238395.1 | - | 10 | 1183 | ||
AK223491.1 | - | 10 | 1270 | |||
AK291832.1 | - | 10 | 1303 | |||
AY927527.1 | - | 3 | 392 | |||
KU178311.1 | - | 10 | 1128 | |||
KU178312.1 | - | 10 | 1128 | |||
U57629.1 | - | 10 | 1187 | |||
X97668.1 | - | 10 | 1128 |