Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEXA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000520.5 | NP_000511.2 | 2 | 816 | ||
NM_001318825.1 | NP_001305754.1 | 2 | 849 | |||
NR_134869.1 | 2 | 816 | ||||
GenBank mRNA | AK222502.1 | - | 2 | 329 | ||
AK296528.1 | - | 2 | 187 | |||
AK297678.1 | - | 3 | 406 | |||
AK301000.1 | - | 2 | 369 | |||
AK301138.1 | - | 2 | 346 | |||
AK307770.1 | - | 2 | 336 | |||
BC018927.2 | - | 2 | 326 | 3353424 | ||
BC084537.1 | - | 2 | 322 | 5459166 | ||
BP356349.1 | - | 2 | 405 | |||
BX537989.1 | - | 2 | 3822 | |||
CR627386.1 | - | 2 | 816 | |||
DC356933.1 | - | 2 | 522 | |||
KU178109.1 | - | 2 | 315 | |||
KU178110.1 | - | 2 | 315 | |||
KU178111.1 | - | 2 | 315 | |||
KU178112.1 | - | 2 | 315 | |||
M13520.1 | - | 2 | 199 |