Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004698.2 | NP_004689.1 | 6, 7 | 898 | ||
XM_011510129.1 | XP_011508431.1 | 7, 8 | 940 | |||
XM_011510130.2 | XP_011508432.1 | 3, 4 | 1460 | |||
XM_011510131.2 | XP_011508433.1 | 6, 7 | 838 | |||
XM_011510132.2 | XP_011508434.1 | 6, 7 | 838 | |||
XM_017002790.1 | XP_016858279.1 | 3, 4 | 334 | |||
XM_017002791.1 | XP_016858280.1 | 6, 7 | 838 | |||
GenBank mRNA | AF001947.1 | - | 5, 6 | 794 | ||
AF016370.1 | - | 6, 7 | 808 | |||
AK222825.1 | - | 6, 7 | 824 | |||
AK297139.1 | - | 6, 7 | 782 | |||
AK297265.1 | - | 5, 6 | 677 | |||
AK299980.1 | - | 7, 8 | 920 | |||
AK310241.1 | - | 3, 4 | 338 | |||
AK311653.1 | - | 7, 8 | 923 | |||
AK312891.1 | - | 6, 7 | 819 | |||
BC000184.2 | - | 5, 6 | 785 | 3351519 | ||
BC001954.1 | - | 5, 6 | 793 | 2905308 |