Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH3A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000382.2 | NP_000373.1 | 3, 4 | 700 | ||
NM_001031806.1 | NP_001026976.1 | 3, 4 | 700 | |||
XM_011523732.1 | XP_011522034.1 | 4, 5 | 609 | |||
XM_011523733.1 | XP_011522035.1 | 4, 5 | 646 | |||
XM_017024355.1 | XP_016879844.1 | 3, 4 | 502 | |||
XM_017024356.1 | XP_016879845.1 | 4, 5 | 608 | |||
XM_017024357.1 | XP_016879846.1 | 4, 5 | 609 | |||
XM_017024358.1 | XP_016879847.1 | 4, 5 | 646 | |||
GenBank mRNA | AB208894.1 | - | 3, 4 | 408 | ||
AK292381.1 | - | 3, 4 | 667 | |||
AK315096.1 | - | 3, 4 | 529 | |||
BC002430.2 | - | 3, 4 | 520 | 3346862 | ||
CR457422.1 | - | 3, 4 | 479 | |||
L47162.1 | - | 3, 4 | 642 | |||
U46689.1 | - | 3, 4 | 928 |