Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000061.2 | NP_000052.1 | 17 | 1897 | ||
NM_001287344.1 | NP_001274273.1 | 17 | 2053 | |||
NM_001287345.1 | NP_001274274.1 | 15 | 1534 | |||
GenBank mRNA | AJ888376.1 | - | 4 | 431 | ||
AJ888378.1 | - | 5 | 504 | |||
AK057105.1 | - | 15 | 1488 | |||
AK309851.1 | - | 17 | 2039 | |||
AK314382.1 | - | 17 | 1864 | |||
AM051275.1 | - | 5 | 459 | |||
AM051276.1 | - | 6 | 717 | |||
AM051278.1 | - | 5 | 652 | |||
AM051280.1 | - | 4 | 351 | |||
AM051282.1 | - | 3 | 263 | |||
BC109079.1 | - | 17 | 1829 | 40034287 | ||
BC109080.1 | - | 17 | 1829 | 40034289 | ||
X58957.1 | - | 17 | 1836 |