Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199282.2 | NP_001186211.2 | 46 | 7252 | ||
NM_006726.4 | NP_006717.2 | 47 | 7244 | |||
XM_005263372.3 | XP_005263429.1 | 47 | 7430 | |||
XM_005263373.2 | XP_005263430.1 | 47 | 7472 | |||
XM_005263374.3 | XP_005263431.1 | 46 | 7396 | |||
XM_005263375.3 | XP_005263432.1 | 46 | 7396 | |||
XM_011532434.2 | XP_011530736.1 | 47 | 7429 | |||
XM_017008872.1 | XP_016864361.1 | 46 | 7439 | |||
XM_017008873.1 | XP_016864362.1 | 7 | 1154 | |||
XM_017008874.1 | XP_016864363.1 | 7 | 860 | |||
GenBank mRNA | AB208838.1 | - | 22 | 2925 | ||
AF216648.2 | - | 47 | 7244 | |||
AF467287.1 | - | 46 | 7252 | |||
AK122920.1 | - | 10 | 1228 | |||
CR749633.1 | - | 7 | 1042 | |||
M83822.1 | - | 29 | 4680 |