Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199282.2 | NP_001186211.2 | 57 | 8785 | ||
NM_006726.4 | NP_006717.2 | 58 | 8780 | |||
XM_005263372.3 | XP_005263429.1 | 58 | 8981 | |||
XM_005263373.2 | XP_005263430.1 | 58 | 9023 | |||
XM_005263374.3 | XP_005263431.1 | 57 | 8947 | |||
XM_005263375.3 | XP_005263432.1 | 57 | 8932 | |||
XM_011532434.2 | XP_011530736.1 | 58 | 8965 | |||
XM_017008872.1 | XP_016864361.1 | 57 | 8990 | |||
XM_017008873.1 | XP_016864362.1 | 18 | 2705 | |||
XM_017008874.1 | XP_016864363.1 | 18 | 2411 | |||
GenBank mRNA | AB208838.1 | - | 33 | 4461 | ||
AF216648.2 | - | 58 | 8780 | |||
AF467287.1 | - | 57 | 8785 | |||
AK026483.1 | - | 11 | 1333 | |||
AK122920.1 | - | 21 | 2764 | |||
BC008644.1 | - | 1 | 190 | |||
BC012535.1 | - | 2 | 73 | |||
BT007180.1 | - | 1 | 19 | |||
BX537552.1 | - | 6 | 1798 | |||
CR749633.1 | - | 18 | 2593 | |||
M83822.1 | - | 40 | 6215 |