Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114173.2 | NP_001107645.1 | 1, 2 | 284 | ||
NM_001814.5 | NP_001805.3 | 1, 2 | 284 | |||
NM_148170.4 | NP_680475.1 | 1, 2 | 284 | |||
GenBank mRNA | AF234263.1 | - | 1, 2 | 195 | ||
AF234264.1 | - | 1, 2 | 195 | |||
AF254757.1 | - | 1, 2 | 195 | |||
AF525032.1 | - | 1, 2 | 195 | |||
AF525033.1 | - | 1, 2 | 195 | |||
AK223038.1 | - | 1, 2 | 233 | |||
AK292117.1 | - | 1, 2 | 202 | |||
AK296190.1 | - | 2, 3 | 182 | |||
AK311923.1 | - | 1, 2 | 233 | |||
BC054028.1 | - | 1, 2 | 223 | |||
BC100891.1 | - | 1, 2 | 219 | |||
BC100892.1 | - | 1, 2 | 219 | |||
BC100893.1 | - | 1, 2 | 219 | |||
BC100894.1 | - | 1, 2 | 219 | |||
BC109386.1 | - | 1, 2 | 219 | |||
BC110071.1 | - | 1, 2 | 219 | |||
BC113850.1 | - | 1, 2 | 219 | |||
BC113897.1 | - | 1, 2 | 219 | |||
BX380095.2 | - | 1, 2 | 284 | |||
BX459226.2 | - | 1, 2 | 268 | |||
BX537913.1 | - | 1, 2 | 258 | |||
CB114425.1 | - | 1, 2 | 265 | |||
X87212.1 | - | 1, 2 | 202 |