Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FGFR1 SIRNA Assays ›
Gene Symbol:
FGFR1
PLAG1
Gene Name:
fibroblast growth factor receptor 1
PLAG1 zinc finger
Gene Aliases:
BFGFR, CD331, CEK, ECCL, FGFBR, FGFR-1, FLG, FLT-2, FLT2, HBGFR, HH2, HRTFDS, KAL2, N-SAM, OGD, bFGF-R-1
PSA, SGPA, ZNF912
Chromosome Location:
Chr.8: 38411138 - 38468834 on Build GRCh38
Chromosome Location:
Chr.8: 56160909 - 56211327 on Build GRCh38
UniGene ID:
Hs.264887
Hs.14968
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
s531163
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FGFR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001174063.1
NP_001167534.1
2
880
NM_001174064.1
NP_001167535.1
2
880
NM_001174065.1
NP_001167536.1
2
200
NM_001174066.1
NP_001167537.1
2
200
NM_001174067.1
NP_001167538.1
3
360
NM_015850.3
NP_056934.2
2
880
NM_023105.2
NP_075593.1
2
880
NM_023106.2
NP_075594.1
2
880
NM_023110.2
NP_075598.2
2
880
XM_006716303.2
XP_006716366.1
2
681
XM_006716304.1
XP_006716367.1
2
197
XM_006716306.2
XP_006716369.1
2
681
XM_006716307.1
XP_006716370.1
2
197
XM_006716310.2
XP_006716373.1
2
681
XM_006716311.1
XP_006716374.1
2
79
XM_006716312.1
XP_006716375.1
2
149
XM_006716313.2
XP_006716376.1
2
681
XM_006716314.1
XP_006716377.1
2
159
XM_011544443.2
XP_011542745.1
2
186
XM_011544444.1
XP_011542746.1
2
181
XM_011544445.2
XP_011542747.1
2
194
XM_011544446.2
XP_011542748.1
2
181
XM_011544447.2
XP_011542749.1
2
181
XM_011544448.1
XP_011542750.1
2
52
XM_011544449.1
XP_011542751.1
2
52
XM_011544450.2
XP_011542752.1
2
52
XM_011544452.2
XP_011542754.1
2
275
XM_017013219.1
XP_016868708.1
2
188
XM_017013220.1
XP_016868709.1
2
181
XM_017013221.1
XP_016868710.1
3
390
XM_017013222.1
XP_016868711.1
2
681
XM_017013223.1
XP_016868712.1
2
197
XM_017013224.1
XP_016868713.1
2
681
XM_017013225.1
XP_016868714.1
2
164
XM_017013226.1
XP_016868715.1
2
52
XM_017013227.1
XP_016868716.1
2
52
XM_017013228.1
XP_016868717.1
2
681
XM_017013231.1
XP_016868720.1
2
279
GenBank mRNA
AB208919.1
-
2
667
AK222718.1
-
2
679
AK223126.1
-
2
681
AK291754.1
-
2
79
AK292470.1
-
2
200
AK309947.1
-
3
239
BC015035.1
-
3
608
3911101
BC018128.1
-
2
644
3896359
BC091494.1
-
1
17
30387634
EF525171.1
-
2
47
FJ809916.1
-
2
880
FJ809917.1
-
2
880
M34185.1
-
2
193
M34186.1
-
2
464
M34187.1
-
2
354
M34188.1
-
2
460
M37722.1
-
2
166
M60485.1
-
3
609
X57119.1
-
2
100
X57120.1
-
2
100
X57121.1
-
2
100
X57122.1
-
2
100
X66945.1
-
2
664
Gene Symbol: PLAG1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
EF525170.1
-
46
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
MAPK cascade
skeletal system development
angiogenesis
ureteric bud development
in utero embryonic development
organ induction
neuron migration
positive regulation of mesenchymal cell proliferation
chondrocyte differentiation
transcription, DNA-templated
protein phosphorylation
sensory perception of sound
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
positive regulation of phospholipase activity
positive regulation of phospholipase C activity
positive regulation of neuron projection development
regulation of phosphatidylinositol 3-kinase signaling
positive regulation of phosphatidylinositol 3-kinase signaling
cell migration
peptidyl-tyrosine phosphorylation
stem cell population maintenance
motogenic signaling involved in postnatal olfactory bulb interneuron migration
ventricular zone neuroblast division
embryonic limb morphogenesis
midbrain development
neuron projection development
fibroblast growth factor receptor signaling pathway involved in orbitofrontal cortex development
phosphatidylinositol-3-phosphate biosynthetic process
inner ear morphogenesis
outer ear morphogenesis
middle ear morphogenesis
chordate embryonic development
positive regulation of MAP kinase activity
positive regulation of MAPK cascade
positive regulation of GTPase activity
regulation of cell differentiation
positive regulation of neuron differentiation
negative regulation of osteoblast differentiation
positive regulation of cell cycle
protein autophosphorylation
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
paraxial mesoderm development
regulation of lateral mesodermal cell fate specification
cell maturation
skeletal system morphogenesis
mesenchymal cell differentiation
regulation of sensory perception of pain
positive regulation of cardiac muscle cell proliferation
auditory receptor cell development
branching involved in salivary gland morphogenesis
lung-associated mesenchyme development
regulation of branching involved in salivary gland morphogenesis by mesenchymal-epithelial signaling
regulation of stem cell proliferation
positive regulation of MAPKKK cascade by fibroblast growth factor receptor signaling pathway
positive regulation of endothelial cell chemotaxis to fibroblast growth factor
regulation of extrinsic apoptotic signaling pathway in absence of ligand
glycoprotein binding
protein tyrosine kinase activity
fibroblast growth factor-activated receptor activity
Ras guanyl-nucleotide exchange factor activity
protein binding
ATP binding
heparin binding
1-phosphatidylinositol-3-kinase activity
fibroblast growth factor binding
protein complex binding
identical protein binding
protein homodimerization activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
cell adhesion molecule binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6c48f4b768-29k6b:80/10.239.166.46:80.
git-commit: a2f5e26549390d543f5d2d1fbad8092df65fbda4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.10.0-2024.06.23-1.0