Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HINT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005340.6 | NP_005331.1 | 1 | 195 | ||
NR_024610.2 | 1 | 195 | ||||
NR_024611.2 | 1 | 195 | ||||
NR_073488.1 | 1 | 195 | ||||
NR_134494.1 | 1 | 195 | ||||
NR_134495.1 | 1 | 195 | ||||
XM_017009410.1 | XP_016864899.1 | 1 | 188 | |||
GenBank mRNA | AK026557.1 | - | 1 | 104 | ||
BC001287.1 | - | 1 | 74 | 3458182 | ||
BC007090.1 | - | 1 | 147 | 4250172 | ||
BF967059.1 | - | 1 | 336 | |||
BG393782.1 | - | 1 | 87 | |||
BG527380.1 | - | 1 | 101 | |||
BG708765.1 | - | 1 | 200 | |||
BG942577.1 | - | 1 | 187 | |||
CD639104.1 | - | 1 | 70 | |||
CK002748.1 | - | 1 | 183 | |||
CN290233.1 | - | 1 | 46 | |||
CN290239.1 | - | 1 | 172 | |||
CR457048.1 | - | 1 | 52 | |||
DB125655.1 | - | 1 | 182 | |||
DB289264.1 | - | 1 | 189 | |||
U27143.1 | - | 1 | 95 | |||
U51004.1 | - | 1 | 159 |