Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMOD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002023.4 | NP_002014.2 | 2 | 535 | ||
GenBank mRNA | AK092899.1 | - | 2 | 205 | ||
AK172740.1 | - | 228 | ||||
AK291632.1 | - | 2 | 201 | |||
AK303866.1 | - | 2 | 201 | |||
AL542756.3 | - | 2 | 598 | |||
BC035281.1 | - | 2 | 212 | 5181410 | ||
X75546.1 | - | 1 | 151 |