Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006265.2 | NP_006256.1 | 4, 5 | 657 | ||
GenBank mRNA | AK289505.1 | - | 4, 5 | 434 | ||
BC050381.1 | - | 4, 5 | 497 | 6044010 | ||
D38551.1 | - | 4, 5 | 553 | |||
X98294.1 | - | 3, 4 | 369 |