Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277961.1 | NP_001264890.1 | 7 | 1446 | ||
NM_001277962.1 | NP_001264891.1 | 7 | 1446 | |||
NM_003156.3 | NP_003147.2 | 7 | 1446 | |||
GenBank mRNA | AK310510.1 | - | 4 | 572 | ||
AK314928.1 | - | 7 | 967 | |||
BC021300.2 | - | 7 | 1438 | 4899542 | ||
HQ317451.1 | - | 7 | 1445 | |||
JX014264.1 | - | 7 | 878 | |||
U52426.1 | - | 7 | 1443 |