Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022336.3 | NP_071731.1 | 8, 9 | 1167 | ||
XM_006712204.1 | XP_006712267.1 | 7, 8 | 1147 | |||
XM_011510502.2 | XP_011508804.2 | 7, 8 | 1026 | |||
XM_011510503.2 | XP_011508805.2 | 8, 9 | 923 | |||
GenBank mRNA | AF130988.1 | - | 8, 9 | 1168 | ||
AK296936.1 | - | 7, 8 | 1057 | |||
AK313781.1 | - | 8, 9 | 1051 | |||
BC093870.1 | - | 7, 8 | 759 | 7939715 | ||
BC093872.1 | - | 7, 8 | 759 | 7939717 | ||
CR749352.1 | - | 8, 9 | 1276 |