Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMCHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015295.2 | NP_056110.2 | 24 | 3186 | ||
XM_011525642.1 | XP_011523944.1 | 24 | 3186 | |||
XM_011525643.2 | XP_011523945.1 | 24 | 3186 | |||
XM_011525644.2 | XP_011523946.1 | 23 | 3094 | |||
XM_017025684.1 | XP_016881173.1 | 25 | 3364 | |||
GenBank mRNA | AK126324.1 | - | 9 | 817 | ||
AW503637.1 | - | 4 | 343 | |||
BU509173.1 | - | 5 | 452 | |||
CR627458.1 | - | 13 | 1410 | |||
DA283721.1 | - | 2 | 138 | |||
DA675822.1 | - | 1 | 41 |